We have updated the storage temperature of the purification columns in this product. For better long-term performance, it is recommended to store the purification columns at 2°C to 8°C.
GeneJET Stabilized and Fresh Whole Blood RNA Kit
Thermo Scientific™

GeneJET Stabilized and Fresh Whole Blood RNA Kit

Thermo Scientific GeneJET™ Stabilized and Fresh Whole Blood RNA Kit is designed for rapid and efficient purification of high qualityRead more
Have Questions?
Catalog NumberQuantity
K087150 preps
Catalog number K0871
Price (USD)
411.00
Each
Quantity:
50 preps
Price (USD)
411.00
Each

Thermo Scientific GeneJET™ Stabilized and Fresh Whole Blood RNA Kit is designed for rapid and efficient purification of high quality total RNA from various whole blood samples: fresh, frozen, or human blood samples stabilized with commercial stabilizers such as Tempus™ and PAXgene™.

The kit utilizes silica-based membrane technology in the form of a convenient spin column, eliminating the need for expensive resins, toxic phenol-chloroform extractions, or time-consuming alcohol precipitation. The kit includes a DNase l for the enzymatic removal of genomic DNA. RNA can be isolated from whole blood in less than 30 minutes (including cell lysis and DNase l treatment).

Highlights

  • Versatile—compatible with fresh and frozen blood, buffy coat, as well as blood collected in PAXgene™and Tempus™ Tubes
  • Convenient—specially optimized Lysis Buffer is suitable for RNA stabilization up to 48 h
  • Efficient—up to 4 μg of total RNA from 500 μL of healthy human blood
  • Pure—isolates intact total RNA free of any RT-qPCR inhibitors

Applications

Isolated total RNA can be used in:

  • RT-PCR
  • RT-qPCR
  • Northern Blotting

Includes

  • Lysis Buffer (R2841)
  • Wash Buffer 1 (conc.) (R2851)
  • Wash Buffer 2 (conc.) (R2861)
  • DNase l (lyophilized)
  • DNase I Reconstitution Buffer
  • 2x DNase I Buffer
  • Manganese Chloride Solution
  • Water, nuclease-free
  • GeneJET RNA Purification Columns pre-assembled with Collection Tubes
  • Collection Tubes, 2 mL
  • Collection Tubes, 1.5 mL

Related Products

Lysis Buffer for GeneJET Blood RNA Kit

Wash Buffer 1 for GeneJET Blood RNA Kit (concentrated)

Wash Buffer 2 for GeneJET Blood RNA Kit (concentrated)

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration2X
For Use With (Application)Real-Time Quantitative PCR (qPCR), Reverse Transcriptase PCR (RT-PCR), Nuclease Protection Assays, Nucleic Acid Labeling, Northern Blotting, cDNA Library Construction
High-throughput CompatibilityNot High-throughput Compatible (Manual)
No. of Reactions50 Preps
Product LineGeneJET
Purification Time30 min.
Quantity50 preps
FormatSpin Column
Isolation TechnologySpin Column
Sample TypeBlood
TargetTotal RNA
Unit SizeEach
Contents & Storage
For better long-term performance, it is recommended to store the purification columns at 2°C-8°C.

Frequently asked questions (FAQs)

Can I use water instead of Elution Buffer in the elution step of GeneJET and MagJET nucleic acid purification kits?

We would highly recommend using the provided Elution Buffer in the elution step for best results. You may use nuclease-free water; however, it may affect the yield of the nucleic acids during purification and their stability during storage.

Is any GeneJET kit suitable for RNA purification from formalin-fixed paraffin-embedded FFPE tissues?

We cannot recommend GeneJET kits for RNA purification from FFPE tissues. In case of genomic DNA purififcation from formalin-fixed paraffin-embedded samples, we recommend GeneJET FFPE DNA Purification Kit.

Are MagJET and GeneJET blood nucleic acid purification kits compatible with animal blood?

Yes, the kits are compatible with animal blood for DNA/RNA purification.

Can I use a vacuum manifold with GeneJET kits?

The GeneJET spin columns are compatible with vacuum manifolds of standard size (e.g. Qiagen, Sigma-Aldrich, Promega). However, not all GeneJET kits are supported with a vacuum protocol. Please refer to the particular kit protocol for the detailed procedure.

At what temperature should I perform centrifugation steps in the GeneJET kits?

We recommend performing centrifugation steps at room temperature when the temperature is not specified.