GeneJET FFPE DNA Purification Kit
Thermo Scientific™

GeneJET FFPE DNA Purification Kit

Thermo Scientific™ GeneJET FFPE DNA Purification Kit is designed for fast and convenient purification of DNA from various amounts of archived FFPE tissue.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
K088150 Preps
K0882250 Preps
Catalog number K0881
Price (USD)
285.65
Online exclusive
311.00
Save 25.35
Each
Quantity:
50 Preps
Price (USD)
285.65
Online exclusive
311.00
Save 25.35
Each

Thermo Scientific™ GeneJET FFPE DNA Purification Kit is designed for fast and convenient purification of DNA from various amounts of archived FFPE tissue. Up to 8 sections (10 μm thickness) of FFPE sample from various tissues can be used for genomic DNA extraction without overnight incubation. Elimination of toxic reagents commonly used for deparaffinization (i.e., xylene) allows a safe DNA purification procedure with high DNA yields and excellent quality. Each preparation recovers up to 8 μg of genomic DNA that can be eluted in volumes from 20 μL allowing for concentrated samples. Due to high decrosslinking efficiency high quality purified DNA shows reliable performance in sensitive downstream applications such as qPCR, PCR (amplicons up to 1 kb) or NGS library preparation.

Highlights

  • High quality DNA – suitable for PCR and qPCR
  • Fast – procedure can be completed within 2 hours
  • Safe – non toxic solvents
  • Efficient – yields up to 8 μg of purified DNA

Applications

  • PCR
  • qPCR
  • NGS library preparation

Includes

  • Proteinase K Solution
  • RNase A Solution
  • Digestion Buffer
  • Binding Buffer
  • Wash Buffer I (conc.)
  • Wash Buffer II (conc.)
  • Elution Buffer
  • GeneJET DNA Purification Columns & Collection Tubes
  • Collection Tubes
  • Detailed Protocol

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Column TypeSpin Column
For Use With (Application)Next-Generation Sequencing, Real-Time Quantitative PCR (qPCR), PCR
High-throughput CompatibilityNot High-throughput Compatible (Manual)
No. of Reactions50 Preps
Product LineGeneJET
Product TypeFFPE DNA Purification Kit
Quantity50 Preps
Sample TypeFFPE, Fixed Samples
TargetGenomic DNA
Test Time<20 min.
FormatSpin Column
Isolation TechnologySpin Column
Unit SizeEach

Frequently asked questions (FAQs)

Is DNA purified from GeneJET FFPE DNA Purification Kit suitable for Next Generation Sequencing (NGS)?

We recommend to evaluate purity with NanoDrop and measure quantity of dsDNA with Qubit$reg; before NGS library preparation. Results can vary depending on the FFPE samples used.

Will quality of DNA purified with GeneJET FFPE DNA Purification Kit affect downstream applications?

Due to high de-crosslinking efficiency of GeneJET FFPE DNA Purification Kit, high-quality purified DNA can be directly used in sensitive downstream applications such as qPCR, PCR (amplicons up to 1 kb) or stored at -20 degrees C.

What are recommended storage conditions for FFPE samples before using them with GeneJET FFPE DNA Purification Kit?

Paraffin sections can be stored at or below +4 degrees C for 1 year without observable negative effects on DNA yield and usability. Longer-term storage of FFPE sections may have negative effects on the DNA due to oxidation.

What are expected yields from the GeneJET FFPE DNA Purification Kit?

DNA yield and quality from FFPE tissue may vary considerably depending on the tissue source, the thickness of the slice, how much tissue is in the slice, the age of the sample, post-sampling delay before fixation, fixation time, etc. We recover up to 8 µg of genomic DNA from one slice (10 µm thickness) of FFPE sample depending on the tissue type.

What amount of FFPE sample can be used with the GeneJET FFPE DNA Purification Kit?

Up to 8 sections (up to 10 µm thickness) of FFPE tissue samples can be used for DNA isolation.