GeneSwitch™ Mammalian Expression Kit, core kit - FAQs
GeneSwitch™ Mammalian Expression Kit, core kit - FAQs
View additional product information for GeneSwitch™ Mammalian Expression Kit, core kit - FAQs (K106002)
10 product FAQs found
Do you offer any GeneSwitch cell lines?
Sorry, all GeneSwitch cell lines have been discontinued.
What are the characteristic features of the GeneSwitch protein?
The GeneSwitch protein contains functional domains from different transcription factors, allowing it to function as a ligand-dependent transcription factor to activate expression of both the gene of interest and its own gene. The GeneSwitch protein exhibits the following characteristics:
- Since the GAL4 DBD is derived from a yeast protein, the GeneSwitch protein has no effect on endogenous genes and can only activate transcription of genes whose expression is controlled by a GAL4 UAS (i.e., the gene of interest and the regulatory fusion gene).
- The GAL4 DBD binds to an individual 17-nucleotide GAL4-binding site as a homodimer. The pGene/V5-His and pSwitch plasmids contain 6 and 4 copies of the GAL4 binding site, respectively, although it is not known if all of the GAL4-binding sites are occupied at any given time.
- The truncated hPR-LBD contains a 19 amino acid deletion from its C-terminal end that abolishes its ability to bind to progesterone, other endogenous steroid hormones, or other progesterone agonists, but still enables it to bind with high affinity to mifepristone.
- The p65 AD is a strong transcriptional activator but is derived from a human protein, to minimize possible toxic or pleiotropic effects associated with viral transactivation domains.
Why is sequential transfection recommended over co-transfection in the T-REx and GeneSwitch systems?
When a co-transfection is performed, there is no way of testing the double stable cell line for functional TetR or GeneSwitch protein, respectively. On the other hand, when sequential transfection is performed, one can functionally test the generated T-REx or GeneSwitch cell line by transiently transfecting the lacZ expression control plasmid and then picking a clone that shows the lowest basal level of expression of lacZ in the absence of the inducer, and the highest level of lacZ in the presence of the inducer. This clone can then be expanded and used to transfect the T-REx or GeneSwitch expression construct, as the case may be.
What is the main advantage of the GeneSwitch system over the T-REx system? And what is its main disadvantage?
With the GeneSwitch system, it is possible to have the absolute lowest basal levels of expression of the gene of interest, whereas the T-REx system may be a little leaky due to the inevitable presence of tetracycline in FBS. The induced level of expression in the GeneSwitch system can be even higher than that seen with the CMV promoter. The disadvantage of the GeneSwitch system is that the expression does not appear to switch off very easily in culture, although it has been demonstrated to function beautifully in transgenics. The T-REx system, on the other hand, can be switched on and off by the addition and removal of the inducer.
I am interested in a mammalian expression system where I can have regulated expression of my gene of interest. I see that you offer multiple systems for this purpose. Can you describe the main features of each system?
We offer three unique mammalian expression systems for inducible/regulated expression of the gene of interest:
- T-REx system
- Flp-In T-REx system
- GeneSwitch system
Please see below to see how they compare with one another:
System -- Basal Expression Level -- Induced Expression Level -- Response time to Maximal Expression -- Transgenic Appliation
T-Rex system -- Low -- Highest -- High -- Suitable
Flp-In T-REx system -- Lower -- High -- 24-48 hrs -- Suitable
GeneSwitch system -- Lowest -- High -- 24-48 hrs -- Suitable
Do I need to include a consensus Kozak sequence when I clone my gene of interest into one of your mammalian expression vectors?
The consensus Kozak sequence is A/G NNATGG, where the ATG indicates the initiation codon. Point mutations in the nucleotides surrounding the ATG have been shown to modulate translation efficiency. Although we make a general recommendation to include a Kozak consensus sequence, the necessity depends on the gene of interest and often, the ATG alone may be sufficient for efficient translation initiation. The best advice is to keep the native start site found in the cDNA unless one knows that it is not functionally ideal. If concerned about expression, it is advisable to test two constructs, one with the native start site and the other with a consensus Kozak. In general, all expression vectors that have an N-terminal fusion will already have an initiation site for translation.
Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.
I sequenced one of your vectors after PCR amplification and observed a difference from what is provided online (or in the manual). Should I be concerned?
Our vectors have not been completely sequenced. Your sequence data may differ when compared to what is provided. Known mutations that do not affect the function of the vector are annotated in public databases.
Are your vectors routinely sequenced?
No, our vectors are not routinely sequenced. Quality control and release criteria utilize other methods.
How was the reference sequence for your vectors created?
Sequences provided for our vectors have been compiled from information in sequence databases, published sequences, and other sources.
What is the consensus Kozak sequence and what is the function of the Kozak sequence?
Eukaryotic (and specifically mammalian) mRNA contains sequence information that is important for efficient translation. However, this sequence, termed a Kozak sequence, is not a true ribosome binding site, but rather a translation initiation enhancer. The Kozak consensus sequence is ACCAUGG, where AUG is the initiation codon. A purine (A/G) in position -3 has a dominant effect; with a pyrimidine (C/T) in position -3, translation becomes more sensitive to changes in positions -1, -2, and +4. Expression levels can be reduced up to 95% when the -3 position is changed from a purine to pyrimidine. The +4 position has less influence on expression levels where approximately 50% reduction is seen. See the following references:
Kozak, M. (1986) Point mutations define a sequence flanking the AUG initiator codon that modulates translation by eukaryotic ribosomes. Cell 44, 283-292.
Kozak, M. (1987) At least six nucleotides preceding the AUG initiator codon enhance translation in mammalian cells. J. Mol. Biol. 196, 947-950.
Kozak, M. (1987) An analysis of 5´-noncoding sequences from 699 vertebrate messenger RNAs. Nucleic Acids Res. 15, 8125-8148.
Kozak, M. (1989) The scanning model for translation: An update. J. Cell Biol. 108, 229-241.
Kozak, M. (1990) Evaluation of the fidelity of initiation of translation in reticulocyte lysates from commercial sources. Nucleic Acids Res. 18, 2828.
Note: The optimal Kozak sequence for Drosophila differs slightly, and yeast do not follow this rule at all. See the following references:
Romanos, M.A., Scorer, C.A., Clare, J.J. (1992) Foreign gene expression in yeast: a review. Yeast 8, 423-488.
Cavaneer, D.R. (1987) Comparison of the consensus sequence flanking translational start sites in Drosophila and vertebrates. Nucleic Acids Res. 15, 1353-1361.
Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.