DreamTaq PCR Master Mix (2X), 20 x 1.25 mL - FAQs
DreamTaq PCR Master Mix (2X), 20 x 1.25 mL - FAQs
View additional product information for DreamTaq PCR Master Mixes (2X) - FAQs (K1072, K1071, K1082, K1081)
2 product FAQs found
What is the error rate of DreamTaq DNA Polymerases?
The error rate of DreamTaq DNA Polymerase is the same as that of Taq DNA Polymerase: 2.2x10^-5 errors per nt per cycle.
Can I do PCR directly from bacterial colonies with Thermo Scientific Taq Master Mixes?
Dream Taq PCR Master Mix and Taq PCR Master Mix can be used for direct PCR from bacterial colonies. Simply scrape a tiny amount of a single colony and transfer to a PCR-tube, prefilled with the master mix. Always use at least 3 min at 95 degrees C for initial denaturation.