GeneBLAzer® AVPR2 CRE-bla CHO-K1 Cell Line -"DISCONTINUED" - FAQs
GeneBLAzer® AVPR2 CRE-bla CHO-K1 Cell Line -"DISCONTINUED" - FAQs
View additional product information for GeneBLAzer® AVPR2 CRE-bla CHO-K1 Cell Line -"DISCONTINUED" - FAQs (K1087)
1 product FAQs found
Is the sequence of the CMV promoter in pcDNA3.1 vector complete, or is it only the core CMV promoter?
In addition to the CAAT and TATA boxes, the CMV promoter in pcDNA3.1 vector contains sequence homologous to base pairs 137 to 724 of the sequence submitted by Boshart, et al (GenBank Accession # K03104). The complete enhancer region is contained between 214 and 620 of this sequence. Therefore, by this definition, the CMV promoter could be said to be "complete".
Please note that this promoter does not contain an intron. Some people believe that the complete promoter must contain the intron, but that has not been demonstrated to be necessary for expression.