InsTAclone PCR Cloning Kit - FAQs
InsTAclone PCR Cloning Kit - FAQs
View additional product information for InsTAclone PCR Cloning Kit - FAQs (K1214)
2 product FAQs found
How long is the control PCR fragment supplied in the InsTAclone PCR Cloning kits (Cat. Nos. K1213, K1214)?
The control PCR fragment is 953 bp long.
What concentration is the vector pTZ57R/T supplied at in the InsTAclone PCR Cloning kits?
The vector pTZ57R/T is supplied at 55 ng/µL.