EpiJET Bisulfite Conversion Kit
Thermo Scientific™

EpiJET Bisulfite Conversion Kit

The EpiJET Bisulfite Conversion Kit is used for an efficient bisulfite modification of DNA for methylation analysis.
Have Questions?
Catalog NumberQuantity
K146150 reactions
Catalog number K1461
Price (ZAR)
-
Quantity:
50 reactions

Thermo Scientific EpiJET Bisulfite Conversion Kit is designed for simple and reliable bisulfite conversion of DNA for methylation analysis. In the bisulfite reaction all unmethylated cytosines are deaminated and converted to uracils, while methylated cytosines remain unchanged. Bisulfite converted unmethylated cytosines are detected as thymines in the following PCR.

The EpiJET Bisulfite Conversion Kit integrates the thermal DNA denaturation and the bisulfite conversion reactions into one step. This step can be performed following long or short protocol, depending on the downstream application of modified DNA. The converted DNA is when bound to a membrane of a micro column for on-column desulfonation and subsequent DNA purification steps. The converted DNA is eluted in a small volume of Elution Buffer and is suitable for a number of techniques used for the methylation status analysis, including PCR, qPCR, COBRA, sequencing.

Highlights

  • High conversion efficiency and specificity (≥99%)
  • Flexibility, provided by short DNA conversion protocol
  • Pure DNA, suitable for downstream applications and long-term storage

Applications

Converted DNA can be used in:

  • PCR
  • qPCR
  • Combined Bisulfite Restriction Analysis (COBRA)
  • Sequencing

Includes

  • Modification Reagent
  • Modification Solution I
  • Modification Solution II
  • Binding Buffer for Bisulfite Conversion Kit
  • Wash buffer (conc.) for Bisulfite Conversion Kit
  • Desulfonation Buffer (conc.)
  • Elution Buffer
  • DNA Purification Micro Columns and Collection Tubes
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product TypeBisulfite Conversion Kit
Quantity50 reactions
Product LineEpiJET
Unit SizeEach

Frequently asked questions (FAQs)

My PCR post-bisulfite conversion failed. What can you suggest?

Here are some recommendations:

- Primers: Ensure that your primers are designed to amplify the converted template. We recommend primers that are 24-32 nts in length and contain no more than 2-3 mixed bases (for base-pairing to C or T residues). The 3' end of your primer should not contain a mixed base or end in a residue whose conversion state is unknown.
- Polymerase: We recommend using a hot-start Taq polymerase such as Platinum Taq DNA Polymerase, Platinum Taq High Fidelity, or AccuPrime Taq DNA Polymerase. Proof-reading polymerases are not recommended because they cannot read through uracil present in DNA templates.
- Amplicon size: Bisulfite modification is harsh and may cause strand breaks. Most research publications recommend 200 bp lengths. Larger amplicons can be generated, but this requires an optimized protocol.
- Template DNA: We recommend using 2-4 µl of eluted DNA for each PCR reaction. Ensure that total template DNA is less than 500 ng.

Where can I find troubleshooting tips for Sanger sequencing of my bisulfite converted DNA?

Please use the following link (http://www.thermofisher.com/us/en/home/technical-resources/technical-reference-library/capillary-electrophoresis-applications-support-center/sanger-sequencing-support/sanger-sequencing-support-troubleshooting.html) for troubleshooting help.

Where can I find troubleshooting tips for next-generation sequencing of my bisulfite converted DNA?

For next-generation sequencing troubleshooting help, please visit the troubleshooting pages within our Next-Generation Sequencing Support Center (thermofisher.com/ngssupport).

I think that my DNA may be under-bisulfite converted (incomplete). What do you recommend?

Ensure that the DNA used for bisulfite conversion is pure. If particulate matter is present after adding the CT Conversion Reagent, then centrifuge the material at high speed and perform the conversion with the clear supernatant. Also, ensure that all of the liquid is at the bottom and not in the cap or walls of the PCR tube before performing the conversion reaction.

What program can I use to design primers for methylation mapping experiments?

You can use Methyl Primer Express Software to design primers for methylation studies. You can download the program for free from here (http://resource.thermofisher.com/page/WE28396_1/).