GeneBLAzer® NPSR1-B-NFAT-bla CHO-K1 Cells - FAQs
GeneBLAzer® NPSR1-B-NFAT-bla CHO-K1 Cells - FAQs
View additional product information for GeneBLAzer® NPSR1-B-NFAT-bla CHO-K1 Cells - FAQs (K1481)
1 product FAQs found
What is the difference between EpiJET 5-hmC Analysis Kit (Cat. No. K1481) and EpiJET 5-hmC and 5-mC Analysis Kit (Cat. No. K1501)?
The new EpiJET 5-hmC and 5-mC Analysis Kit is more universal than the previous EpiJET 5-hmC Analysis Kit because it provides a total assessment of both cytosine modifications - 5-hydroxymethylcytosine (5-hmC) and methylcytosine (5-mC) - within specific DNA loci (for comparison - the current kit allows only 5-hmC analysis). Additionally, all necessary DNA controls do not originate from plasmids widely used in molecular biology laboratories are included into the new kit.