PureLink™ Genomic Plant DNA Purification Kit - FAQs
PureLink™ Genomic Plant DNA Purification Kit - FAQs
View additional product information for PureLink™ Genomic Plant DNA Purification Kit - FAQs (K183001)
1 product FAQs found
I'm interested in isolating plant DNA, but am worried about the abundance of carbohydrates and complex polysaccharides in the tissue. What kit do you recommend for this?
We offer several kits specifically designed to isolate plant DNA depending on the amount of starting material and chemistry you are interested in using. Please use this link for a comparison of our kits available for plant DNA extraction.