S.N.A.P.™ Gel Purification Kit - FAQs
S.N.A.P.™ Gel Purification Kit - FAQs
View additional product information for S.N.A.P.™ Gel Purification Kit - FAQs (K199925)
6 product FAQs found
I'm running an agarose gel with TAE or TBE. Can I use your gel extraction kits?
Yes, both TAE and TBE agarose gels are compatible with our gel extraction kits.
What is the difference between Purelink PureLink Quick Gel Extraction System and S.N.A.P. Gel Purification Kit?
The PureLink kit can be used with both TAE and TBE gels, while the S.N.A.P. Gel Purification Kit is recommended for TAE gels only.
Can you use TBE gels with S.N.A.P. gel purification kits?
No, TBE gels contain borate, which interferes with the sodium iodide solution. Therefore, only TAE gels can be used. We recommend the PureLink Quick Gel Extraction kit for both TAE and TBE agarose gels.
What is the smallest fragment that can be purified with S.N.A.P. columns?
S.N.A.P. mini prep columns have been used to purify fragments of 100 bp in gel purification procedures.
What is the resin in the S.N.A.P. kits?
The column contains a silica/glass fiber based resin. However, the exact composition is proprietary.
What is unique about the S.N.A.P. UV-free Gel Purification Kit?
The S.N.A.P. UV-free Gel Purification Kit utilizes crystal violet as a staining reagent, which can be visualized in non-UV light conditions. This can prevent DNA damage that can be caused by the UV light required by normal ethidium bromide staining. DNA fragments recovered using this kit have higher cloning efficiency. In addition, DNA can be visualized under normal light as a thin violet band while the gel is running and excised as soon as bands are sufficiently resolved.