TA Cloning™ Kit, Dual Promoter, with pCR™II Vector and One Shot™ INVαF' Chemically Competent E. coli, 20 reactions - FAQs
TA Cloning™ Kit, Dual Promoter, with pCR™II Vector and One Shot™ INVαF' Chemically Competent E. coli, 20 reactions - FAQs
View additional product information for TA Cloning™ Kit, Dual Promoter, with pCR™II Vector and One Shot™ INVαF' Chemically Competent E. coli - FAQs (K205040, K205001)
1 product FAQs found
Have you compared in vitro transcription levels between SP6 and T7 promoters in your pCRII vectors?
No, we have not done in-house comparisons of transcription levels. It is widely known though that T7 polymerase produces more RNA than SP6 (on the order of 10-fold higher).