PureLink™ Nucleic Acid Purification Rack - FAQs
PureLink™ Nucleic Acid Purification Rack - FAQs
View additional product information for PureLink™ Nucleic Acid Purification Rack - FAQs (K210013)
1 product FAQs found
How do I eliminate the RNA contamination I get in my minipreps?
Low copy number plasmids may be contaminated with RNA due to the large number of cells being processed. Increase the concentration of RNAse in the buffers 4 fold.