MagJET Whole Blood gDNA Kit

Catalog number: K2741

Thermo Scientific™  Related applications: DNA Extraction

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Thermo Scientific MagJET Whole Blood Genomic DNA Kit is designed for fast and efficient purification of genomic DNA from fresh or frozen whole blood treated with EDTA or citrate, as well as blood fractions. Note: Samples preserved with heparin are not compatible with this kit.

The kit utilizes paramagnetic bead technology enabling high yields and robust performance. High binding capacity, uniform particle size, and rapid magnetic response of MagJET magnetic beads makes the technology ideal for high throughput automatic nucleic acid purification, as well as for manual purification by low sample throughput users.

The resulting high quality DNA is free of proteins, nucleases and other contaminants or inhibitors, can be used in a wide range of downstream applications such as PCR, qPCR and other enzymatic reactions.

Highlights

Compatible—suitable to use with common stabilizers such as EDTA, citrates
Versatile—suitable for gDNA purification from fresh, frozen or dried blood, as well as bone marrow and buffy coat
Efficient—high yields of pure DNA free of contaminants and downstream application inhibitors
Flexible—easily adjustable for high throughput automation platforms and manual processing

Applications

• Isolated genomic DNA is ideal for use in common molecular biology procedures, including:
• PCR and qPCR
• FastDigest and conventional restriction digestion
• Southern blotting
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Sample Type (General): Blood
Downstream Application: Cloning, PCR, Real-Time Quantitative PCR (qPCR), Sequencing, Southern Blotting
High Throughput Compatibility: High Throughput-Compatible, Not High Throughput-Compatible (Manual)
Product Line: MagJET
Product Size: 96 preps

Documents

Manuals & protocols