MagJET Plant Genomic DNA Kit

Catalog number: K2762

Thermo Scientific™  Related applications: DNA Extraction

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Thermo Scientific MagJET Plant Genomic DNA Kit is designed for automated high throughput or manual purification of genomic DNA from a wide variety of plant species. The kit exploits an efficient magnetic particle technology eliminating need for phenol/chloroform extraction or alcohol precipitation.

The MagJET Plant Genomic DNA Kit procedures are optimized for the isolation of genomic DNA from 10-100 mg plant samples. DNA yields vary between different species and tissues depending on genome size, ploidy, cell number, and age of tissue sample.

The kit utilizes paramagnetic bead technology enabling high yields and robust performance. High binding capacity, uniform particle size, and rapid magnetic response of MagJET magnetic beads makes the technology ideal for high throughput automatic nucleic acid purification, as well as for manual purification for low sample throughput.

The resulting high quality DNA is free of proteins, nucleases and other contaminants or inhibitors, and can be used in a wide range of downstream applications such as PCR, qPCR or other enzymatic reactions. See Table 1 for typical genomic DNA yields from various sources.

Highlights

Versatile—suitable for any plant type and part, including woody, polyphenol-rich and lignified samples
High gDNA yields—compared to traditional CTAB method
High quality—isolates high integrity gDNA, free of inhibitors and RNA contamination
Flexible—easily adjustable for high throughput automation platforms and manual processing

Applications

• Isolated genomic DNA is ideal for use in common molecular biology procedures, including:
• PCR and qPCR FastDigest and conventional restriction digestion
• Southern blotting
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Sample Type (General): Plant Samples
Downstream Application: Cloning, PCR, Real-Time Quantitative PCR (qPCR), Sequencing, Southern Blotting
High Throughput Compatibility: High Throughput-Compatible, Not High Throughput-Compatible (Manual)
Product Line: MagJET
Product Size: 4 x 96 preps

Documents

Manuals & protocols