RapidOut DNA Removal Kit - FAQs
RapidOut DNA Removal Kit - FAQs
View additional product information for RapidOut DNA Removal Kit - FAQs (K2981)
2 product FAQs found
Does DRR (DNase Binding Reagent) from RapidOut DNA Removal Kit bind to RNA present in enzymatic reaction mixtures? What is the recovery of RNA after the procedure?
RNA recovery after the RapidOut DNA removal procedure is high. Typically it is higher than conventional DNAse I inactivation methods such as heat, EDTA or chloroform/phenol extraction.
Does the DRR (DNase Binding Reagent) from RapidOut DNA Removal Kit bind to other proteins? Is the kit suitable for removal of other proteins from the reaction mixture?
We have tested the kit for DNase I removal only. Although DRR can bind to other proteins, we cannot provide any recommendations for removal of protein contaminants from the enzymatic mixtures. If the RNA sample contains a considerable amount of protein contaminants, modify the RNA purification protocol to reduce protein contamination.