PureLink™ PCR Micro Kit
Invitrogen™

PureLink™ PCR Micro Kit

PureLink PCR Micro Kits are ideal for purifying and concentrating DNA from low-yielding PCR or restriction digestion reactions. These highlyRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
K310250250 Preps
K31005050 Preps
Catalog number K310250
Price (USD)
786.00
Each
Quantity:
250 Preps
Price (USD)
786.00
Each

PureLink PCR Micro Kits are ideal for purifying and concentrating DNA from low-yielding PCR or restriction digestion reactions. These highly purified DNA fragments and amplicons provide excellent results in downstream applications such as DNA sequencing, restriction digestion, cloning, PCR, and labeling. PureLink PCR Micro Kits provide a simple and rapid method for purifying and concentrating DNA fragments ranging in size from 100 bp-12 kb.

The unique micro-nozzle design allows up to 95% recovery of DNA fragments with as little as a 5 μL elution volume. With less than a 10% coefficient of variance in the elution volume you can be sure that no ethanol is carried over from the wash steps giving you the best chance of success in downstream applications. Removal of primers, dNTPs, enzymes, and salts exceeds 98.5%. This kit can also efficiently remove unincorporated dye-labeled nucleotides from a cDNA labeling reaction.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Elution Volume Max20 μL
Elution Volume Min5 μL
FormatSpin Column
High-throughput CompatibilityNot High-throughput Compatible (Manual)
Isolation TechnologySilica Spin Column
No. of Reactions250 Preps
Product LinePureLink
Product TypePCR Micro Kit
Quantity250 Preps
Run Time10 min.
Sample TypePCR Products
Separation Range100 bp to 12 kb
Shipping ConditionRoom Temperature
Starting Material Volume125 μL
TargetDNA from Enzymatic Reactions (e.g. PCR)
For Use With (Application)Cloning, Standard PCR
Unit SizeEach
Contents & Storage
Kit includes columns, binding and wash buffers, recovery tubes, and the manual. Store at room temperature.

Frequently asked questions (FAQs)

What are the size limitations for the PureLink PCR Cleanup Micro Kit?

The size cut-off is 100 bp to 12 kb.

What are the minimum and maximum size ranges for DNA fragment cleanup using the PureLink PCR Clean-Up Kit?

The size range is between 100 bp and 12 kb. Our kit comes with two buffers, Binding Buffer (B2) and Binding Buffer HC (B3). The Binding Buffer HC (B3) eliminates primer-dimers and short failed PCR products that are smaller than 300 bp.