IMAGEN™ Human Metapneumovirus (hMPV) Kit using Direct Immunofluorescence Assay
Thermo Scientific™
IMAGEN™ Human Metapneumovirus (hMPV) Kit using Direct Immunofluorescence Assay
Carry out rapid detection of hMPV in clinical samples with this direct immunofluorescence assay.
Have Questions?
Catalog NumberQuantity
K612511-250 Tests
Catalog number K612511-2
Price (USD)
-
Quantity:
50 Tests
Rapidly detect hMPV in clinical samples with Thermo Scientific™ IMAGEN™ Human Metapneumovirus (hMPV) Kit. This direct immunofluorescence assay utilizes two monoclonal antibodies in order to detect specific structural proteins expressed in all strains of human metapneumovirus.
Human metapneumovirus (hMPV) is a respiratory virus that causes an upper respiratory infection. It causes seasonal epidemics centred around the winter months, and symptoms include fever with upper respiratory symptoms such as cough and nasal congestion1, 2. However, hMPV has also been associated with more severe illness and lower respiratory tract symptoms such as pneumonia and bronchiolitis3,4.
Use IMAGEN Human metapneumovirus (hMPV) Kit as a rapid, sensitive and specific method for direct detection of hMPV in clinical samples such as nasopharyngeal aspirates.
  • Rapid results- within little over 30 minutes of receiving test sample or culture
  • Simple- ready-to-use reagents plus all IMAGEN assays follow the same basic methodology
  • Excellent sensitivity and specificity- high quality, well-characterized antibodies used in the assay provide excellent sensitivity and specificity.
  • Easy-to-read results
  • Long Shelf Life- typically 24 months.
Human metapneumovirus (hMPV) is an enveloped, spherical RNA virus of the family Paramyxoviridae, subfamily Paramyxovirinae and is classified within the genus Metapneumovirus3. hMPV is the only member of the genus that affects humans, with the other members including avian pneumoviruses A, B, C and D1,5.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific™ brand.
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
General References:
  1. Bastien N., Ward D., Van Caeseel P., Brandt K., Lee S.H.S., McNabb G., Klisko B., Chan E., Li Y. (2003) Human Metapneumovirus in the Canadian Population.Journal of Clinical Microbiology 41: 4642–4646
  2. Hamelin M-E., Abed Y., Boivin G. (2004) Human Metapneumovirus: A New Player among Respiratory Viruses. Emerging Infections 38: 983–990.1. Frankl R.I.B.,
  3. Fauquet C.M., Knudson D.L., Brown F. (1992) Classification and Nomenclature of Viruses. Fifth Report of the International Committee on Taxonomy of Viruses. Archives of Virology Supplement 2, Spurger Velacy, New York, pp 245–246.
  4. Easton A.J., Domachowske J.B., Rosenburg H.F. (2004) Animal Pneumoviruses: Molecular genetics and Pathogenesis.Clinical Microbiology Reviews Apr. 200, p. 390-412
  5. van den Hoogen B.G., de Jong J.C., Groen J., Kuiken T., de grootR., Frouchier R.A., Osterhaus A.D. (2001) A Newly Discovered Human Pneumovirus Isolated from Young Children with Respiratory Tract Disease. Nature Medicine 7: 719-724
Specifications
DescriptionIMAGEN Human metapneumovirus Kit
Includes2 x 1 positive control slide, mounting fluid, hMPV Reagent, instruction for use
Quantity50 Tests
Detectable AnalytesHuman Metapneumovirus (hMPV) Antigen
Unit SizeEach
Contents & Storage
2°C to 8°C