How does expression in MagicMedia affect the solubility of proteins when compared to standard LB and IPTG induction?
In most cases, the ratio of soluble to insoluble proteins is not much different when compared to LB and IPTG. However, since more total protein is typically obtained in MagicMedia, one also obtains more total soluble protein.