MagicMedia™ E. coli Expression Medium, Powder, 5 x 1 L - FAQs
MagicMedia™ E. coli Expression Medium, Powder, 5 x 1 L - FAQs
View additional product information for MagicMedia™ E. coli Expression Medium, Powder - FAQs (K6810, K6815)
3 product FAQs found
Can I use a lower incubation temperature for MagicMedia Medium?
Yes, expression at 18 degrees C and 25 degrees C has been validated using the Dual Temperature expression protocol.
Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.
I am using a Champion pET vector with the BL21-AI cells. Can I use your MagicMedia Medium for expression?
Unfortunately, MagicMedia Medium will not work with the arabinose inducer. MagicMedia Medium needs an IPTG-inducible system along with an intact Lac operon.
Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.
What is MagicMedia Medium?
MagicMedia Medium is an E. coli expression medium. It produces 3-10 times higher protein yield, is compatible with T7 regulated E. coli vectors that contain a functional lac operon and BL21 (DE3) strains, is simple to use, and eliminates OD monitoring/induction steps.
Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.