ProBond™ Purification System with Antibody -"DISCONTINUED" - FAQs
ProBond™ Purification System with Antibody -"DISCONTINUED" - FAQs
View additional product information for ProBond™ Purification System with Antibody -"DISCONTINUED" - FAQs (K85301)
1 product FAQs found
What expression levels can be expected with the pTrcHis/CAT construct?
In one experiment, 35 µg CAT/mg total protein or 68.2 mg CAT/liter of culture was obtained. 50 ml of cell extract was loaded onto a ProBond column and 2 mg of CAT was recovered. The eluted protein appeared reasonably pure on a gel.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.