본 제품은 액체 질소로 배송 및 보관되는 세포 관련 제품으로, 해당 제품 구매 시 세포 사용 가이드라인을 반드시 확인해주십시오. 세포사용가이드라인 다운로드
Product Image
Gibco™
FreeStyle™ MAX CHO Expression System
The FreeStyle MAX System is a complete system for mammalian transient protein production. This system combines Gibco FreeStyle media, FreeStyle자세히 알아보기
Have Questions?
카탈로그 번호수량
K9000201 kit
카탈로그 번호 K900020
제품 가격(KRW)
2,132,000
온라인 행사
Ends: 30-Jun-2026
2,508,000
할인액 376,000 (15%)
Each
수량:
1 kit
제품 가격(KRW)
2,132,000
온라인 행사
Ends: 30-Jun-2026
2,508,000
할인액 376,000 (15%)
Each
The FreeStyle MAX System is a complete system for mammalian transient protein production. This system combines Gibco FreeStyle media, FreeStyle MAX Reagent, and either FreeStyle CHO-S cells or FreeStyle 293 cells. The only equipment required to use the FreeStyle MAX System is a CO2 incubator and a platform shaker with shake flasks or a spinner culture.
The FreeStyle MAX System provides:
• An easy, rapid protocol for transient transfection of CHO and 293 cells in suspension cultures
• Up to milligram quantities of functional proteins with the expected post-translational modifications
• Simplified downstream purification of secreted proteins with FreeStyle™ serum-free medium
• Straightforward protocol for large-scale production
The FreeStyle MAX System can replace time-consuming stable cell line generation with large-scale functional protein production in one week.
For Research Use Only. Not for use in diagnostic procedures.
사양
세포주FreeStyle™ CHO-S Cells
분류Animal Origin-Free, Serum-Free
배양 유형Suspension Cell Culture
배달 유형Transfection
발현 메커니즘Cell-Based Expression
발현 시스템Mammalian
용도(애플리케이션)Protein Expression
주요 기능Protein Production
제품라인Freestyle
제품 유형Expression System
수량1 kit
셀 유형CHO
Transfection TechniqueLipid-based Transfection
Unit SizeEach
구성 및 보관
Store FreeStyle cells in liquid nitrogen. Store FreeStyle MAX Reagent at 2–8°C. Store FreeStyle CHO Expression Medium, FreeStyle 293 Expression Medium, and OptiPRO SFM at 2–8°C in the dark. Store the pCMV SPORT at -20°C. Stability is guaranteed for 6 months under the recommended storage conditions. Please refer to the manual for additional storage information.
자주 묻는 질문(FAQ)
I am interested in a mammalian expression system that will allow me to obtain very high yields of my protein. What kind of systems do you offer?
For stable high-yield expression, we offer the Freedom CHO-S and Freedom DG44 kits. For transient high-yield expression, we offer the ExpiCHO Expression System, Expi293 Expression System, FreeStyle 293 Expression System, FreeStyle Max 293 Expression System, and FreeStyle Max CHO Expression System. For high-yield expression of functional membrane proteins in Exp293F cells, we offer the Expi293 MembranePro Expression System (Cat. Nos. A25869, A25870) that combines the scalability and ease of use of Expi293 and the technology of MembranePro to allow an increase of more than 20-fold in membrane protein yield compared to the standard, adherent culture MembranePro Functional Protein Expression System. Please see the Application Note (http://www.thermofisher.com/content/dam/LifeTech/global/life-sciences/ProteinExpressionAnalysis/pdfs/PG1391-PJ6257-CO28704-Expi293Adaption-MembraneProTech_AppNoteGlobal-FHR.pdf) for more details.
Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.
What vector do you recommend using with FreeSytle MAX CHO Expression System (Cat. No. K900020)?
We recommend using the pcDNA 3.4 TOPO TA Cloning Kit (Cat. No. A14697).
Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.