PicoPure™ DNA Extraction Kit - FAQs
PicoPure™ DNA Extraction Kit - FAQs
View additional product information for PicoPure™ DNA Extraction Kit - FAQs (KIT0103)
2 product FAQs found
What is the time requirment for DNA extraction when using the PicoPure DNA Extraction kit?
The total incubation time depends on the tissue source for DNA extraction. Non-formalin fixed samples are more efficient, taking approximately 3 hours. Formalin-fixed tissues require more time due to cross-linking, taking approxiately 16 hours or more.
What kit do you recommend using for DNA isolation from Arcturus Laser Capture Microdissection (LCM) samples?
The Arcturus PicoPure DNA Extraction Kit (Cat. No. KIT0103) offers an easy, streamlined genomic extraction procedure that produces PCR-ready DNA. It allows you to extract and amplify DNA in the same tube, without organic extraction or spin columns, and to recover DNA from as few as 10 cells prepared by laser capture microdissection (LCM) or other tissues. The kit provides conveniently packaged, stable Proteinase K, PCR-compatible DNA Reconstitution Buffer and a complete user guide.