How many cells are needed for the PicoPure RNA Isolation Kit?
Find additional tips, troubleshooting help, and resources within our Nucleic Acid Purification and Analysis Support Center.
Search
View additional product information for PicoPure™ RNA Isolation Kit - FAQs (KIT0214, KIT0204)
7 product FAQs found
In addition, a typical Bioanalyzer profile of RNA from an FFPE sample can look fragmented and degraded and as a result, that approach may not be useful in determining the transcribability of your RNA. Therefore, a tissue scrape test is recommended. You can use the entire adjacent section to isolate RNA and to access the RNA quantity and quality for intact RNA. Two sets of β-actin gene-specific primers designed to amplify short segments of DNA within the β-actin target sequence from the 5’ and 3’ of the β-actin gene can be used to estimate the amount of RNA in a given sample in relation to β-actin content using real-time PCR. The RNA quantity derived from the 3' primer set is used to quantitate the RNA in the FFPE tissue scrape. The ratio of the RNA yield obtained from both sets of PCR primers is the 3'⁄5' ratio and is used as an indication of RNA quality. If there is less product yield of the 5’ β-actin primers, it indicates that there is less full length of RNA.