Product Image
Applied Biosystems™
Arcturus™ Paradise™ Plus RNA Extraction and Isolation Kit
The ARCTURUS™ Paradise™ PLUS FFPE RNA Isolation Kit permits unprecedented gene expression analysis of millions of previously inaccessible samples byMás información
Have Questions?
Número de catálogoCantidad
KIT0312I12 muestras
Número de catálogo KIT0312I
Precio (CLP)
-
Cantidad:
12 muestras
The ARCTURUS™ Paradise™ PLUS FFPE RNA Isolation Kit permits unprecedented gene expression analysis of millions of previously inaccessible samples by unlocking RNA from formalin-fixed, paraffin-embedded (FFPE) tissues. Amplify as little as 5 ng of the isolated fixed RNA using reagents and methods that have been specially optimized to overcome the challenges of cross-linked FFPE templates (Figure 1).
Secure Downstream Success with the Paradise™ QC Kit
Not all FFPE samples contain high quality RNA. Identify the best samples for your studies using a simple, real-time PCR research technique, and then proceed with confidence using these pre-qualified samples (Figure 2).
Integrated Systems for Microgenomics
The Paradise™ PLUS Reagent System is validated as part of the ARCTURUS™ complete System for Microgenomics™, an integrated solution for utilizing small quantities of RNA for gene expression analysis, and features the ARCTURUSXT™ Laser Capture Microdissection (LCM) Instrument to procure pure cell populations. ARCTURUS™' Systems for Microgenomics enable accurate and sensitive microarray assays that reveal molecular otherwise obscured in whole tissue samples or cell mixtures.
For Research Use Only. Not for use in diagnostics procedures.
Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.
Especificaciones
Tipo de producto finalARN total
Para utilizar con (aplicación)Análisis de micromatrices, PCR en tiempo real cuantitativa (qPCR)
Para utilizar con (equipo)ArcturusXT™ LCM Instrument
Compatibilidad de alto rendimientoNo compatible con alto rendimiento (manual)
N.º de muestras12 samples
Línea de productosARCTURUS, Paradise
Cantidad12 muestras
Cantidad de material de partidaAs little as a single cell.
FormatoSpin Column
Isolation TechnologyEnzymatic, Spin Column
Tipo de muestraLaser Capture Microdissection (LCM), Tissue (FFPE)
TargetTotal RNA
Unit SizeEach
Contenido y almacenamiento
Paradise RNA Isolation Kit: Proteinase K, Protenaise K buffer, purification reagents, purification columns, microcentrifuge tubes. Variable storage conditions
Preguntas frecuentes
What is the lowest amount of RNA that can be amplified using the Arcturus Paradise Plus RNA Extraction and Isolation Kit?
With this kit, you can amplify as little as 5 ng of RNA isolated from formalin-fixed, paraffin embedded (FFPE) tissues.