Related applications: Laser Capture Microdissection
Error loading your content!
Catalog number |
Select a plan
Unit size
|
Price ({{currency}}) | Availability | Qty | |
---|---|---|---|---|---|
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} |
|
— |
Downstream Application: | Microarray Analysis, Nucleic Acid Labeling |
---|---|
For Use With (Equipment): | ArcturusXT™ LCM Instrument |
Product Line: | ARCTURUS™, Paradise® |
Product Size: | 1 kit |
Sample Type (General): | FFPE & Fixed Samples |
Sample Type (Specific): | Tissue (FFPE) |
Workflow Step: | In Vitro Transcription, RNA Amplification, RNA Extraction, Reverse Transcription, Tissue Staining |