RNA amplification using the Van Gelder and Eberwine technique (Van Gelder 1990) utilizes an oligo(dT) primer containing the T7 RNA polymerase promoter for synthesis of first strand cDNA. The poly(A) tail at the end of mRNA sequences serves as the substrate for the binding of these primers. Since mRNA typically constitutes only 1-5% of the total RNA in the cell, only this fraction of the total RNA is amplified. The tissue type, its developmental state, and its health all influence the actual proportion of mRNA in a total RNA sample. Total RNA from brain, testes, and embryonic tissues may contain up to 4% mRNA, while RNA from many other tissues will have only 1% or less mRNA. The RNA isolation method can also influence mRNA content. The generally accepted average value for mRNA content is about 2% of a total RNA sample. When 1 µg of total RNA, 2% or 20 ng of which is mRNA, is amplified 1000-fold, yields of 20 µg aRNA (or cRNA) should be expected. You may observe higher fold amplification when starting with lower amounts of total RNA. This is because, in an in vitro transcription (IVT) reaction, a finite amount of RNA can be synthesized with the fixed amount of NTPs. When starting with less RNA, NTPs do not become limiting until the RNA is amplified beyond the typical 1000-2000 fold amplification levels seen with higher amounts of input RNA.
Find additional tips, troubleshooting help, and resources within our
Epigenetics Support Center.