Arcturus™ RiboAmp™ PLUS Kit, 6 Samples - FAQs
Arcturus™ RiboAmp™ PLUS Kit, 6 Samples - FAQs
View additional product information for Arcturus™ RiboAmp™ PLUS Kit - FAQs (KIT0521)
6 product FAQs found
What is the typical size range of amplified RNA?
A single round of amplification yields product sizes ranging from 200 bases to 6 kb. The majority of these products are approximately 1.5 kb in length. A second round of amplification will result in shorter products. We recommend using an Agilent 2100 bioanalyzer to visualize these products. Amplification products can be visualized by agarose gel electrophoresis; they will migrate as a smear. Although this data is still useful, it is less informative than bioanalyzer analysis.
Find additional tips, troubleshooting help, and resources within our Epigenetics Support Center.
Why is RNA amplification necessary?
Glass microarray analysis experiments typically require 5-20 µg of total RNA per slide for sample labeling and hybridization. Thus, microarray-based gene expression analysis of very small samples [laser capture microdissection (LCM), tissue biopsies, or other clinical samples] is difficult due to the very low amounts of total RNA recovered from the samples. Linear amplification of RNA from small samples produces sufficient quantities of RNA for sample labeling and hybridization. Since the amplification technique is highly reproducible and maintains representation of the gene expression in the original sample, it is recommended for probe synthesis by most manufacturers of commercially available microarrays.
Find additional tips, troubleshooting help, and resources within our Epigenetics Support Center.
How is fold amplification calculated?
RNA amplification using the Van Gelder and Eberwine technique (Van Gelder 1990) utilizes an oligo(dT) primer containing the T7 RNA polymerase promoter for synthesis of first strand cDNA. The poly(A) tail at the end of mRNA sequences serves as the substrate for the binding of these primers. Since mRNA typically constitutes only 1-5% of the total RNA in the cell, only this fraction of the total RNA is amplified. The tissue type, its developmental state, and its health all influence the actual proportion of mRNA in a total RNA sample. Total RNA from brain, testes, and embryonic tissues may contain up to 4% mRNA, while RNA from many other tissues will have only 1% or less mRNA. The RNA isolation method can also influence mRNA content. The generally accepted average value for mRNA content is about 2% of a total RNA sample. When 1 µg of total RNA, 2% or 20 ng of which is mRNA, is amplified 1000-fold, yields of 20 µg aRNA (or cRNA) should be expected. You may observe higher fold amplification when starting with lower amounts of total RNA. This is because, in an in vitro transcription (IVT) reaction, a finite amount of RNA can be synthesized with the fixed amount of NTPs. When starting with less RNA, NTPs do not become limiting until the RNA is amplified beyond the typical 1000-2000 fold amplification levels seen with higher amounts of input RNA.
Find additional tips, troubleshooting help, and resources within our Epigenetics Support Center.
How do I locate the overview image on the Arcturus XT LCM Instrument?
First, optimize the visualization of the image in the live video at 2X by increasing or decreasing the brightness setting, then right-click on the overview image and select "remember settings". This resets your optimized image settings, and when you select "re-acquire overview image" you will now generate a perfectly exposed overview image.
With the ArcturusXT LCM Instrument, which laser should I use to isolate my region of interest? UV cutting laser or IR capture laser?
For a few cells: The gentle IR-only approach is the best recommendation for preserving nucleic acids and verifying that the desired material is collected on the cap. For groups of cells or large tumor regions: Mount your tissue on a membrane slide and take advantage of the power of the IR, or IR + UV cutting laser to cut around your region of interest quickly and cleanly.
What is Arcturus Laser Capture Microdissection (LCM)?
The ArcturusXT LCM Instrument has a proprietary combination of a gentle IR laser and a powerful UV laser that work in conjunction to efficiently isolate cells from frozen sections or paraffin embedded sections without changing morphology or integrity of the biological content. The IR laser helps to capture the cells of interest, and the UV laser microdissects the captured cells. This is done without affecting the morphology of the cells, and allows for visual inspection of the remaining specimen to help ensure the quality of the sample collected. In addition to the flexibility that the dual laser system provides, there is the flexibility of multiple stage inserts for various sample types. A wide-slide stage format is available for neurobiology researchers working with very large sections of brain tissue. The petri dish stage insert enables live-cell microdissection applications such as stem cell research and other rare-cell isolations.