Search
View additional product information for Mouse Amyloid beta 42 ELISA Kit - FAQs (KMB3441)
2 product FAQs found
At the present time, we offer ELISA kits for the quantitation of beta amyloid 1-40 and beta amyloid 1-42 specifically. The rabbit polyclonal detection antibody directed to the C-terminus of beta amyloid is the component which imparts the specificity to the assay. For the kits for measuring beta amyloid 1-40, we use a rabbit polyclonal which is specific for the beta amyloid 40 cleavage site as the detection antibody. This antibody allows the kit to discern beta amyloid 40 from beta amyloid 42 and beta amyloid 43. For the kits for measuring beta amyloid 42, we use a rabbit polyclonal antibody which is specific for the beta amyloid 42 cleavage site as the detection antibody. This antibody allows the kit to discern beta amyloid 42 from beta amyloid 40 and beta amyloid 43.
Yes, the detection antibody is specific to the C-terminus of Amyloid beta 42.