IL-12 p70 Mouse ELISA Kit

Catalog number: KMC0121C

Invitrogen™  Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The IL-12 Mouse ELISA Kit quantifies natural and recombinant human IL-12 heterodimer and the free p40 unit in serum, buffered solution, or cell culture medium. IL-12 is a lymphokine.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Gene Symbol: IL12A, IL12B
Assay Incubation Time: 3 hr
Assay Range: 7.8-500 pg/mL
Assay Sensitivity: <2 pg/mL
Concentration: 7.8-500 pg⁄ml
Detection Method: Colorimetric
For Use With (Equipment): Microplate Reader
Gene Aliases: IL-12p35, Il-12a, Ll12a, p35, Il-12b, Il-12p40, Il12p40, p40
Gene ID: 16159, 16160
Gene Name: interleukin 12A, interleukin 12b
Immunoassay Kit Format: Coated ELISA Kit
Label or Dye: HRP (Horseradish Peroxidase)
Marker: IL-12
Product Line: Novex™
Product Size: 480 assays
Protein Aliases: IL-12A, Cytotoxic lymphocyte maturation factor 35 kDa subunit, CLMF p35, IL-12 subunit p35, IL-12B, Cytotoxic lymphocyte maturation factor 40 kDa subunit, CLMF p40, IL-12 subunit p40
Protein Name: Interleukin-12 subunit alpha, Interleukin-12 subunit beta
Sample Type (Specific): Cell Culture Supernatants, Serum
Sample Volume: 50 µl
Shipping Condition: Wet Ice
Species: Mouse
Target (Kit Named): IL-12 p70
Target Species (Named): Mouse
Target Species (Validated): Mouse
UniProt ID: P43431, P43432

Contents & storage

Pre-coated plate(s), standard or calibrator, detector antibody, HRP conjugate, diluents, wash buffer, substrate(s), and stop solution. Store kit at 2-8°C. See product manual for detailed contents and storage conditions for maximum stability.

Documents

Manuals & protocols

Recommended products