N-(Hydroxymethyl)trifluoroacetamide, 98+%
N-(Hydroxymethyl)trifluoroacetamide, 98+%
N-(Hydroxymethyl)trifluoroacetamide, 98+%
Thermo Scientific Chemicals

N-(Hydroxymethyl)trifluoroacetamide, 98+%

CAS: 50667-69-1 | C3H4F3NO2 | 143.065 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L00128.06
also known as L00128-06
Price (USD)
58.65
Special offer
Online exclusive
Ends: 28-Dec-2026
69.00
Save 10.35
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
58.65
Special offer
Online exclusive
Ends: 28-Dec-2026
69.00
Save 10.35
Each
Chemical Identifiers
CAS50667-69-1
IUPAC Name2,2,2-trifluoro-N-(hydroxymethyl)acetamide
Molecular FormulaC3H4F3NO2
InChI KeyURUWXKFAEKTWKG-UHFFFAOYSA-N
SMILESOCNC(=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Assay (HPLC)≥98.0%
FormCrystalline powder or powder
Melting Point92-108°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only