Butadiene monoxide, 98%
Butadiene monoxide, 98%
Butadiene monoxide, 98%
Thermo Scientific Chemicals

Butadiene monoxide, 98%

CAS: 930-22-3 | C4H6O | 70.091 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number L00238.06
also known as L00238-06
Price (USD)
181.00
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
181.00
Each
Chemical Identifiers
CAS930-22-3
IUPAC Name2-ethenyloxirane
Molecular FormulaC4H6O
InChI KeyGXBYFVGCMPJVJX-UHFFFAOYNA-N
SMILESC=CC1CO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥97.5%
Refractive Index1.4155-1.4200 @ 20?C
Identification (FTIR)Conforms
FormLiquid
Butadiene monoxide is used as key starting material for the synthesis of (-)-bulgecinine and (+)-broussonetine G.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Butadiene monoxide is used as key starting material for the synthesis of (-)-bulgecinine and (+)-broussonetine G.

Solubility
Miscible with ethanol, ethyl ether, benzene and organic solvents.

Notes
Moisture sensitive. Store in cool place. Incompatible with acids, bases and oxidizing agents.
RUO – Research Use Only

General References:

  1. Reaction with allyl bromide, mediated by In metal, affords diallylmethanol: Tetrahedron Lett., 44, 2911 (2003):
  2. Li, G.; Feng, X.; Du, H. Palladium-catalyzed asymmetric allylic amination of racemic butadiene monoxide with isatin derivatives. Org. Biomol. Chem. 2015, 13 (20), 5826-5830.
  3. Mao, W.; He, H. H.; Gu, J. Q.; Chen, W.; Wu, K.; Tok, E. S.; Xu, G. Q. Preservation of epoxy groups on surfaces in the covalent attachment of butadiene monoxide on Si(111)-(7x7): the effect of a vinyl substituent. Chem. Commun. 2015, 51 (75), 14195-14198.