(Ethoxycarbonylmethyl)dimethylsulfonium bromide, 98%
(Ethoxycarbonylmethyl)dimethylsulfonium bromide, 98%
(Ethoxycarbonylmethyl)dimethylsulfonium bromide, 98%
Thermo Scientific Chemicals

(Ethoxycarbonylmethyl)dimethylsulfonium bromide, 98%

CAS: 5187-82-6 | C6H13BrO2S | 229.132 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number L00266.18
also known as L00266-18
Price (USD)
204.65
Online Exclusive
227.00
Save 22.35 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
204.65
Online Exclusive
227.00
Save 22.35 (10%)
Each
Chemical Identifiers
CAS5187-82-6
IUPAC Name(2-ethoxy-2-oxoethyl)dimethylsulfanium bromide
Molecular FormulaC6H13BrO2S
InChI KeyJXFPTJYKYKVENJ-UHFFFAOYSA-M
SMILES[Br-].CCOC(=O)C[S+](C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥97.5%
Appearance (Color)White
Assay (Argentometric Titration)≥97.5 to ≤102.5%
FormCrystals or powder or crystalline powder or fused solid or granules
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only