2-Chloroacrylic acid, 96%, stab. with ca 0.3% BHT
2-Chloroacrylic acid, 96%, stab. with ca 0.3% BHT
2-Chloroacrylic acid, 96%, stab. with ca 0.3% BHT
2-Chloroacrylic acid, 96%, stab. with ca 0.3% BHT
Thermo Scientific Chemicals

2-Chloroacrylic acid, 96%, stab. with ca 0.3% BHT

CAS: 598-79-8 | C3H3ClO2 | 106.51 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
Catalog number L00350.09
also known as L00350-09
Price (USD)
279.65
Online Exclusive
311.00
Save 31.35 (10%)
Each
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)
279.65
Online Exclusive
311.00
Save 31.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS598-79-8
IUPAC Name2-chloroprop-2-enoic acid
Molecular FormulaC3H3ClO2
InChI KeySZTBMYHIYNGYIA-UHFFFAOYSA-N
SMILESOC(=O)C(Cl)=C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥95.0 to ≤105.0%
Appearance (Color)White to cream to brown or pale pink
FormCrystals or powder or crystalline powder or lumps or fused solid
Stabilizer≈0.3% BHT
2-Chloroacrylic acid is used as a halochemicals, organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloroacrylic acid is used as a halochemicals, organic synthesis.

Solubility
Soluble in water.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only

General References:

  1. Paul J. Marsden.; John E. Casida. 2-Haloacrylic acids as indicators of mutagenic 2-haloacrolein intermediates in mammalian metabolism of selected promutagens and carcinogens. J. Agric. Food Chem. 1982, 30, (4), 627-631.
  2. Atsushi Kurata.; Tatsuo Kurihara.; Harumi Kamachi.; Nobuyoshi Esaki. Asymmetric reduction of 2-chloroacrylic acid to (S)-2-chloropropionic acid by a novel reductase from Burkholderia sp. WS. Tetrahedron: Asymmetry. 2004, 15, (18), 2837-2839.