Thermo Scientific Chemicals

Ethyl diethoxyacetate, 97+%, Thermo Scientific Chemicals

Catalog number: L00390.22
100 g, Each
Thermo Scientific Chemicals

Ethyl diethoxyacetate, 97+%, Thermo Scientific Chemicals

Catalog number: L00390.22
100 g, Each
Quantity
Catalog number: L00390.22
also known as L00390-22
Price (USD)
Price: 201.00
Online price: 171.65
Your price:
Quantity
-

Chemical Identifiers

CAS
6065-82-3
IUPAC Name
ethyl 2,2-diethoxyacetate
Molecular Formula
C8H16O4
InChI Key
XCLBIKIQSCTANZ-UHFFFAOYSA-N
SMILES
CCOC(OCC)C(=O)OCC
Refractive Index
1.4080-1.4130 @ 20?C
Form
Liquid
Appearance (Color)
Clear colorless
Assay (GC)
≥97.0%

Description

Ethyl diethoxyacetate is used in the preparation of a fatty acid analog containing DTS, 12,13-dioxotetradecanoic acid di(N-methyl-thiosemicarbazone) (FA-DTS). It is used for synthesis of heterocyclic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl diethoxyacetate is used in the preparation of a fatty acid analog containing DTS, 12,13-dioxotetradecanoic acid di(N-methyl-thiosemicarbazone) (FA-DTS). It is used for synthesis of heterocyclic compounds.

Solubility
Soluble in Dichloromethane, Ethyl Acetate and Methanol.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text