3'-Methylacetophenone, 97%
3'-Methylacetophenone, 97%
3'-Methylacetophenone, 97%
3'-Methylacetophenone, 97%
Thermo Scientific Chemicals

3'-Methylacetophenone, 97%

CAS: 585-74-0 | C9H10O | 134.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L00440.06
also known as L00440-06
Price (USD)
26.65
Special offer
Online exclusive
Ends: 30-Jun-2026
31.20
Save 4.55 (15%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
26.65
Special offer
Online exclusive
Ends: 30-Jun-2026
31.20
Save 4.55 (15%)
Each
Add to cart
Chemical Identifiers
CAS585-74-0
IUPAC Name1-(3-methylphenyl)ethan-1-one
Molecular FormulaC9H10O
InChI KeyFSPSELPMWGWDRY-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=CC(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to pale yellow to pale brown
FormPowder or clear liquid as melt
Identification (FTIR)Conforms
Assay (GC)≥96.0%
Refractive Index1.5285-1.5345 @ 20?C
3'-Methylacetophenone is used in Pharmaceutical Intermediates and in Organic Intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3′-Methylacetophenone is used in Pharmaceutical Intermediates and in Organic Intermediates.

Solubility
Difficult to mix.

Notes
Protect from heat. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Krishnakumar, V.; Balachandran, V. DFT studies, vibrational spectra and conformational stability of 4-hydroxy-3-methylacetophenone and 4-hydroxy-3-methoxyacetophenone. Spectrochimica Acta Part A: Molecular and Biomolecular Spectroscopy. 2005, 61, (11-12), 2510-2525.
  2. Peter J. Hylands.; Kristin Ingolfsdottir. The isolation of methyl β-orsellinate from Stereocaulon alpinum and comments on the isolation of 4,6-dihydroxy-2- methoxy-3-methylacetophenone from stereocaulon species. Phytochemistry. 1985, 24, (1), 127-129.