tert-Nitrosobutane dimer, 98+%
tert-Nitrosobutane dimer, 98+%
tert-Nitrosobutane dimer, 98+%
tert-Nitrosobutane dimer, 98+%
Thermo Scientific Chemicals

tert-Nitrosobutane dimer, 98+%

CAS: 6841-96-9 | C8H18N2O2 | 174.244 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number L00462.03
also known as L00462-03
Price (USD)
98.80
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
98.80
Each
Add to cart
Chemical Identifiers
CAS6841-96-9
IUPAC Name2-methyl-2-nitrosopropane
Molecular FormulaC4H9NO
InChI KeyLUQZKEZPFQRRRK-UHFFFAOYSA-N
SMILESCC(C)(C)N=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale green or blue
FormCrystals or powder or crystalline powder
Assay (HPLC)≥98.0%
tert-Nitrosobutane dimer is used as a spin trapping reagent in electron spin resonance studies. Spin trap reagent, Reactant involved in the synthesis of air-stable π-radical based on Corannulene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Nitrosobutane dimer is used as a spin trapping reagent in electron spin resonance studies. Spin trap reagent, Reactant involved in the synthesis of air-stable π-radical based on Corannulene.

Solubility
Soluble in 5% chloroform.

Notes
Light Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
Forms adducts, nitroxyl radicals, with free radicals for identification by ESR. For review of spin trapping, see: Acc. Chem. Res., 4, 31 (1971).