2,2-Dimethyl-1,3-dioxolane, 98+%
2,2-Dimethyl-1,3-dioxolane, 98+%
2,2-Dimethyl-1,3-dioxolane, 98+%
Thermo Scientific Chemicals

2,2-Dimethyl-1,3-dioxolane, 98+%

CAS: 2916-31-6 | C5H10O2 | 102.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L00482.14
also known as L00482-14
Price (USD)
97.65
Online Exclusive
108.00
Save 10.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
97.65
Online Exclusive
108.00
Save 10.35 (10%)
Each
Chemical Identifiers
CAS2916-31-6
IUPAC Name2,2-dimethyl-1,3-dioxolane
Molecular FormulaC5H10O2
InChI KeySIJBDWPVNAYVGY-UHFFFAOYSA-N
SMILESCC1(C)OCCO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Refractive Index1.3965-1.4005 @ 20?C
Assay (GC)≥98.0%
2,2-Dimethyl-1,3-dioxolane is used as organic intermediates, fine chemicals, pharmaceutical research and development.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2-Dimethyl-1,3-dioxolane is used as organic intermediates, fine chemicals, pharmaceutical research and development.

Solubility
Insoluble in water.

Notes
Protect from heat. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Field, N.D. The Preparation of 1,3-Dioxole and 2,2-Dimethyl-1,3-dioxole. J. Am. Chem. Soc. 1961, 83, (16), 3504-3507.
  2. Corey, E.J.; Saizo Shibata.; Raman K. Bakshi. An efficient and catalytically enantioselective route to (S)-(-)-phenyloxirane. J. Org. Chem. 1988, 53, (12), 2861-2863.
  3. Reagent for the conversion, by exchange, of carbonyl compounds to their ethylene acetals: J. Am. Chem. Soc., 76, 1359 (1954).