(4-Methoxybenzyl)triphenylphosphonium chloride, 97%
(4-Methoxybenzyl)triphenylphosphonium chloride, 97%
(4-Methoxybenzyl)triphenylphosphonium chloride, 97%
(4-Methoxybenzyl)triphenylphosphonium chloride, 97%
Thermo Scientific Chemicals

(4-Methoxybenzyl)triphenylphosphonium chloride, 97%

CAS: 3462-97-3 | C26H24ClOP | 418.90 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number L00594.09
also known as L00594-09
Price (USD)
77.40
Each
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)
77.40
Each
Add to cart
Chemical Identifiers
CAS3462-97-3
IUPAC Name[(4-methoxyphenyl)methyl]triphenylphosphanium chloride
Molecular FormulaC26H24ClOP
InChI KeyYQXBNCFNXOFWLR-UHFFFAOYSA-M
SMILES[Cl-].COC1=CC=C(C[P+](C2=CC=CC=C2)(C2=CC=CC=C2)C2=CC=CC=C2)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder
Assay (Titration ex Chloride)≥96.0 to ≤104.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only