3-Benzylidene-2,4-pentanedione, 97%
3-Benzylidene-2,4-pentanedione, 97%
3-Benzylidene-2,4-pentanedione, 97%
Thermo Scientific Chemicals

3-Benzylidene-2,4-pentanedione, 97%

CAS: 4335-90-4 | C12H12O2 | 188.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L00623.06
also known as L00623-06
Price (USD)
34.65
Online Exclusive
37.80
Save 3.15 (8%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
34.65
Online Exclusive
37.80
Save 3.15 (8%)
Each
Chemical Identifiers
CAS4335-90-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Refractive Index1.5800-1.5850 @ 20?C
Assay (GC)≥96.0%
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only