N,N-Dimethylformamide di-n-propyl acetal, 97%
N,N-Dimethylformamide di-n-propyl acetal, 97%
N,N-Dimethylformamide di-n-propyl acetal, 97%
Thermo Scientific Chemicals

N,N-Dimethylformamide di-n-propyl acetal, 97%

CAS: 6006-65-1 | C9H22NO2 | 176.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number L00643.09
also known as L00643-09
Price (USD)
51.65
Online exclusive
57.30
Save 5.65
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
51.65
Online exclusive
57.30
Save 5.65
Each
Chemical Identifiers
CAS6006-65-1
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.4070-1.4120 @ 20?C
Appearance (Color)Clear colorless
Assay (GC)≥96.0%
N,N-Dimethylformamide di-n-propyl acetal, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethylformamide di-n-propyl acetal, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
It hydrolyzes with water.

Notes
Moisture, water Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
RUO – Research Use Only

General References:

  1. NC Jain.; DM Chinn.; RD Budd.; TS Sneath. Simultaneous determination of cocaine and benzoyl ecgonine in urine by gas chromatography with on-column alkylation. J. forensic Sci. 197722,(1) , 7-15.
  2. KJ Miller. Additivity methods in molecular polarizability. Journal of the American Chemical Society. 1990112,(23) , 8533-8542.