Phenylglyoxylic acid, 98%
Phenylglyoxylic acid, 98%
Phenylglyoxylic acid, 98%
Thermo Scientific Chemicals

Phenylglyoxylic acid, 98%

CAS: 611-73-4 | C8H6O3 | 150.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L00648.14
also known as L00648-14
Price (USD)
113.65
Special offer
Online exclusive
Ends: 30-Jun-2026
133.00
Save 19.35 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
113.65
Special offer
Online exclusive
Ends: 30-Jun-2026
133.00
Save 19.35 (15%)
Each
Chemical Identifiers
CAS611-73-4
IUPAC Name2-oxo-2-phenylacetic acid
Molecular FormulaC8H6O3
InChI KeyFAQJJMHZNSSFSM-UHFFFAOYSA-N
SMILESOC(=O)C(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Clarity2.5% w/v solution in ethanol will be clear to very faintly turbid by JIS K8001 5.2
Assay (HPLC)≥97.5%
Melting Point (clear melt)60-69?C
FormCrystalline powder
Appearance (Color)White to cream or pale yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only