o-Toluic hydrazide, 98+%
o-Toluic hydrazide, 98+%
o-Toluic hydrazide, 98+%
Thermo Scientific Chemicals

o-Toluic hydrazide, 98+%

CAS: 7658-80-2 | C8H10N2O | 150.181 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L00668.14
also known as L00668-14
Price (USD)
255.65
Online Exclusive
284.00
Save 28.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
255.65
Online Exclusive
284.00
Save 28.35 (10%)
Each
Chemical Identifiers
CAS7658-80-2
IUPAC Name2-methylbenzohydrazide
Molecular FormulaC8H10N2O
InChI KeyKFXLXEQCRFGDRU-UHFFFAOYSA-N
SMILESCC1=CC=CC=C1C(=O)NN
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Melting Point (clear melt)120.0-127.0?C
Appearance (Color)White to pale cream to pale yellow
Assay (GC)≥98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only