(+/-)-2-Phenylpropionic acid, 98%
(+/-)-2-Phenylpropionic acid, 98%
(+/-)-2-Phenylpropionic acid, 98%
Thermo Scientific Chemicals

(+/-)-2-Phenylpropionic acid, 98%

CAS: 492-37-5 | C9H10O2 | 150.177 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number L00709.06
also known as L00709-06
Price (USD)
35.65
Online Exclusive
39.20
Save 3.55 (9%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
35.65
Online Exclusive
39.20
Save 3.55 (9%)
Each
Chemical Identifiers
CAS492-37-5
IUPAC Name2-phenylpropanoic acid
Molecular FormulaC9H10O2
InChI KeyYPGCWEMNNLXISK-UHFFFAOYNA-N
SMILESCC(C(O)=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Silylated GC)>97.5%
FormLiquid
Refractive Index1.5210-1.5250 @ 20?C
Appearance (Color)Clear, pale yellow
Assay (Aqueous acid-base Titration)>97.5 to <102.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only