5,6-Dimethoxyindole-2-carboxylic acid, 98%
5,6-Dimethoxyindole-2-carboxylic acid, 98%
5,6-Dimethoxyindole-2-carboxylic acid, 98%
Thermo Scientific Chemicals

5,6-Dimethoxyindole-2-carboxylic acid, 98%

CAS: 88210-96-2 | C11H11NO4 | 221.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L00733.06
also known as L00733-06
Price (USD)
329.65
Online Exclusive
366.00
Save 36.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
329.65
Online Exclusive
366.00
Save 36.35 (10%)
Each
Chemical Identifiers
CAS88210-96-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to brown or grey to dark grey
FormPowder
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
5,6-Dimethoxyindole-2-carboxylic acid is used as a reagent and heterocyclic building blocks in synthetic chemistry. It is used to prepare a melanin-like polymer, poly(5,6-dimethoxyindole-2-carboxylic acid), which exhibits electrochromic properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5,6-Dimethoxyindole-2-carboxylic acid is used as a reagent and heterocyclic building blocks in synthetic chemistry. It is used to prepare a melanin-like polymer, poly(5,6-dimethoxyindole-2-carboxylic acid), which exhibits electrochromic properties.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Gunbas, G.; Toppare, L. Electrochromic conjugated polyheterocycles and derivatives-highlights from the last decade towards realization of long lived aspirations. Chem. Commun. 2012, 48 (8), 1083-1101.
  2. Kim, Y. J.; Wu, W.; Chun, S. E.; Whitacre, J. F.; Bettinger, C. J. Biologically derived melanin electrodes in aqueous sodium-ion energy storage devices. Proc. Natl. Acad. Sci. U.S.A 2013, 110 (52), 20912-20917.