2-Chloro-2,2-difluoroacetamide, 98%
2-Chloro-2,2-difluoroacetamide, 98%
2-Chloro-2,2-difluoroacetamide, 98%
Thermo Scientific Chemicals

2-Chloro-2,2-difluoroacetamide, 98%

CAS: 354-28-9 | C2H2ClF2NO | 129.491 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L00785.14
also known as L00785-14
Price (USD)
555.65
Special offer
Online exclusive
Ends: 30-Jun-2026
653.00
Save 97.35 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
555.65
Special offer
Online exclusive
Ends: 30-Jun-2026
653.00
Save 97.35 (15%)
Each
Chemical Identifiers
CAS354-28-9
IUPAC Name2-chloro-2,2-difluoroacetamide
Molecular FormulaC2H2ClF2NO
InChI KeyPSAKKOKLSDIKEK-UHFFFAOYSA-N
SMILESNC(=O)C(F)(F)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥97.5%
Water Content (Karl Fischer Titration)≤1%
FormCrystals or powder or crystalline powder or fused solid
Appearance (Color)White to cream to pale brown
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only