5-Hydroxyindole-2-carboxylic acid, 97%
5-Hydroxyindole-2-carboxylic acid, 97%
5-Hydroxyindole-2-carboxylic acid, 97%
5-Hydroxyindole-2-carboxylic acid, 97%
Thermo Scientific Chemicals

5-Hydroxyindole-2-carboxylic acid, 97%

CAS: 21598-06-1 | C9H6NO3 | 176.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L00786.03
also known as L00786-03
Price (USD)
68.65
Online Exclusive
76.00
Save 7.35 (10%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
68.65
Online Exclusive
76.00
Save 7.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS21598-06-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to cream to yellow to pale brown
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Assay (HPLC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only