(Methoxymethyl)diphenylphosphine oxide, 98+%
(Methoxymethyl)diphenylphosphine oxide, 98+%
(Methoxymethyl)diphenylphosphine oxide, 98+%
Thermo Scientific Chemicals

(Methoxymethyl)diphenylphosphine oxide, 98+%

CAS: 4455-77-0 | C14H15O2P | 246.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number L00912.06
also known as L00912-06
Price (USD)
76.65
Special offer
Online exclusive
Ends: 30-Jun-2026
89.70
Save 13.05 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
76.65
Special offer
Online exclusive
Ends: 30-Jun-2026
89.70
Save 13.05 (15%)
Each
Chemical Identifiers
CAS4455-77-0
IUPAC Name[(methoxymethyl)(phenyl)phosphoroso]benzene
Molecular FormulaC14H15O2P
InChI KeyOEPKDBQOTLDTNC-UHFFFAOYSA-N
SMILESCOCP(=O)(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder
Assay (GC)≥98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only