2-Bromobenzhydrazide, 98+%
2-Bromobenzhydrazide, 98+%
2-Bromobenzhydrazide, 98+%
Thermo Scientific Chemicals

2-Bromobenzhydrazide, 98+%

CAS: 29418-67-5 | C7H7BrN2O | 215.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L00937.14
also known as L00937-14
Price (USD)
124.65
Special offer
Online exclusive
Ends: 30-Jun-2026
147.00
Save 22.35 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
124.65
Special offer
Online exclusive
Ends: 30-Jun-2026
147.00
Save 22.35 (15%)
Each
Chemical Identifiers
CAS29418-67-5
IUPAC Name2-bromobenzohydrazide
Molecular FormulaC7H7BrN2O
InChI KeyPQNLAYLOCZKPIY-UHFFFAOYSA-N
SMILESNNC(=O)C1=CC=CC=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)149.5-155.5?C
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder or fused/lumpy solid
Assay (GC)≥98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only