Phenyl carbamate, 98+%
Phenyl carbamate, 98+%
Phenyl carbamate, 98+%
Thermo Scientific Chemicals

Phenyl carbamate, 98+%

CAS: 622-46-8 | C7H7NO2 | 137.138 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L00944.06
also known as L00944-06
Price (USD)
43.65
Online Exclusive
48.30
Save 4.65 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
43.65
Online Exclusive
48.30
Save 4.65 (10%)
Each
Chemical Identifiers
CAS622-46-8
IUPAC Namephenyl carbamate
Molecular FormulaC7H7NO2
InChI KeyBSCCSDNZEIHXOK-UHFFFAOYSA-N
SMILESNC(=O)OC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder or fused solid
Assay (HPLC)≥98.0%
Identification (FTIR)Conforms
Phenyl carbamate, is used as various phenyl carbamate esters derived from amino acids, a dipeptide and amino acid esters and amides were prepared and assessed as potential prodrugs with the aim of protecting phenolic drugs against first-pass metabolism following per oral administration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenyl carbamate, is used as various phenyl carbamate esters derived from amino acids, a dipeptide and amino acid esters and amides were prepared and assessed as potential prodrugs with the aim of protecting phenolic drugs against first-pass metabolism following per oral administration.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
RUO – Research Use Only

General References:

  1. ZH Fu.; Y Ono. Synthesis of methyl N-phenyl carbamate by methoxycarbonylation of aniline with dimethyl carbonate using Pb compounds as catalysts. Journal of molecular catalysis. 199491(3) , 399-405.
  2. RA Coss.; JD Pickett-Heaps. The effects of isopropyl N-phenyl carbamate on the green alga Oedogonium cardiacum I. Cell Division. The Journal of cell biology. 197463(1) , 84-98 .