4-Amino-3-hydrazino-5-mercapto-1,2,4-triazole, 99+%, Thermo Scientific Chemicals
4-Amino-3-hydrazino-5-mercapto-1,2,4-triazole, 99+%, Thermo Scientific Chemicals
4-Amino-3-hydrazino-5-mercapto-1,2,4-triazole, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Amino-3-hydrazino-5-mercapto-1,2,4-triazole, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L00982.14
Price (USD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS1750-12-5
IUPAC Name4-amino-3-hydrazinyl-4,5-dihydro-1H-1,2,4-triazole-5-thione
Molecular FormulaC2H6N6S
InChI KeyRDIMQHBOTMWMJA-UHFFFAOYSA-N
SMILESNNC1=NNC(=S)N1N
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (HPLC)≥99.0%
Identification (FTIR)Conforms
Appearance (Color)White to cream to pale brown to pale gray to gray
4-Amino-3-hydrazino-5-mercapto-1,2,4-triazole is a reagent used in the determination of aldehydes and other reactive chemicals.was used in purpald assay for determination of bacterial polysaccharides containing substituted or unsubstituted glycol in residues such as glycerol, ribitol, arabinitol, furanosyl galactose and sialic acid. It was used as chromogen during the determination of catalase activity in small tissue samples. It was used as reagent in determination of formaldehyde. It was used for derivatization of neutral monosaccharides during HPLC.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Amino-3-hydrazino-5-mercapto-1,2,4-triazole is a reagent used in the determination of aldehydes and other reactive chemicals.was used in purpald assay for determination of bacterial polysaccharides containing substituted or unsubstituted glycol in residues such as glycerol, ribitol, arabinitol, furanosyl galactose and sialic acid. It was used as chromogen during the determination of catalase activity in small tissue samples. It was used as reagent in determination of formaldehyde. It was used for derivatization of neutral monosaccharides during HPLC.

Solubility
Soluble in Dimethyl sulfoxide. (DMSO)

Notes
Store at -20°C. Avoid contact with heat. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Kunihiro KamatA; Misako Takahashi; Kiyoshi Terajima and Motohiro Nishijima. Spectrophotometric determination of sodium chondroitin sulfate in eye drops after derivatization with 4-amino-3-hydrazino-5-mercapto-1,2,4-triazole.Analyst.1995, 120 2755-2758.
  2. M. J. Del Nozala; J. L. Bernala; V. Hernandeza; L. Toribioa & R. Mendezb. Purpald (4-Amino-3-hydrazino-5-mercapto-1, 2, 4-triazole) as a Reagent for Post-Column Derivatization of Neutral Monosaccharides in High Pressure Liquid Chromatography.Journal of Liquid Chromatography.1993, 16 1105-1116.
  3. Reagent for the detection and classification of aldehydes: Chem. Commun., 1719 (1970); J. Chem. Educ., 55, 206 (1978). The reagent gives no color reaction with ketones.